This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SIGIRR SIRNA Assays ›
Gene Symbol:
Sigirr
Gene Name:
single immunoglobulin and toll-interleukin 1 receptor (TIR) domain
Gene Aliases:
AI256711, TIR8
Chromosome Location:
Chr.7: 141091175 - 141100610 on Build GRCm38
UniGene ID:
Mm.38017
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 188835

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sigirr
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023059.3 NP_075546.2 5, 6 1082
GenBank mRNA AF113795.1 - 4, 5 599
AF239957.1 - 4, 5 593
AK133518.1 - 5, 6 707
AK145889.1 - 5, 6 1085
BC010806.1 - 4, 5 604 4240425
BC094069.1 - 5, 6 688 4974962

Back To Top

More Information


Gene Ontology Categories:

Function(s)

negative regulation of cytokine-mediated signaling pathway
acute-phase response
signal transduction
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of chemokine biosynthetic process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0