This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TCEA1 SIRNA Assays ›
Gene Symbol:
Tcea1
Gene Name:
transcription elongation factor A (SII) 1
Gene Aliases:
S-II
Chromosome Location:
Chr.1: 4857694 - 4897909 on Build GRCm38
UniGene ID:
Mm.207263
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 187224

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tcea1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159750.1 NP_001153222.1 10 2318
NM_001159751.1 NP_001153223.1 10 2214
NM_011541.4 NP_035671.1 10 2321
GenBank mRNA AK136035.1 - 10 2214
BC061490.1 - 10 2211 2582185
BC083127.1 - 10 2184 6390364
M18209.1 - 9 2119

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
erythrocyte differentiation
regulation of DNA-templated transcription, elongation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
nucleic acid binding
DNA binding
protein binding
zinc ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0