This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPATCH11 SIRNA Assays ›
Gene Symbol:
Gpatch11
Gene Name:
G patch domain containing 11
Gene Aliases:
2310002B06Rik, C80922, Ccdc75, L26697
Chromosome Location:
Chr.17: 78835516 - 78848308 on Build GRCm38
UniGene ID:
Mm.220253
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 186536

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gpatch11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181649.6 NP_857632.4 4 417
GenBank mRNA AK009084.1 - 4 304
AK148331.1 - 4 419
BC047998.1 - 4 331 6511097
BC092522.1 - 4 344 30648550
BC138310.1 - 4 312 8861330
BY741452.1 - 4 418

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
nucleic acid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0