This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF24 SIRNA Assays ›
Gene Symbol:
Rnf24
Gene Name:
ring finger protein 24
Gene Aliases:
2810473M14Rik, 4930505A13Rik, AI317164, C86507, D2Ertd504e
Chromosome Location:
Chr.2: 131298488 - 131352892 on Build GRCm38
UniGene ID:
Mm.477523
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 186514

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf24
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178607.4 NP_848722.1 6 2597
GenBank mRNA AK048813.1 - 6 2555
AK089207.1 - 6 2584
AK165427.1 - 6 2555
BC096529.1 - 6 2530 6406985

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
zinc ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0