This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP410 SIRNA Assays ›
Gene Symbol:
Zfp410
Gene Name:
zinc finger protein 410
Gene Aliases:
Apa1, D12Ertd748e, Znf410
Chromosome Location:
Chr.12: 84315928 - 84344439 on Build GRCm38
UniGene ID:
Mm.255150
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 186398

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp410
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252582.1 NP_001239511.1 3 658
NM_001252583.1 NP_001239512.1 3 623
NM_144833.3 NP_659082.1 3 623
GenBank mRNA AK033090.1 - 3 400
AK042224.1 - 3 380
AK049384.1 - 4 678
AK053001.1 - 3 348
AK083003.1 - 3 365
AK132345.1 - 3 361
AK139797.1 - 2 177
BC021528.1 - 3 378 5353673
BC052080.1 - 3 355 6403411
CK791111.1 - 3 623

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
nucleic acid binding
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0