This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GLMP SIRNA Assays ›
Gene Symbol:
Glmp
Gene Name:
glycosylated lysosomal membrane protein
Gene Aliases:
0610031J06Rik, AB027141, NCU-G1
Chromosome Location:
Chr.3: 88325023 - 88328631 on Build GRCm38
UniGene ID:
Mm.439922
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 185885

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Glmp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020003.1 NP_064387.1 4 814
GenBank mRNA AB027141.1 - 4 770
AB027142.1 - 4 814
AK007803.1 - 4 787
AK075614.1 - 4 792
AK080538.1 - 4 788
AK150794.1 - 4 787
AK154340.1 - 4 787
AK160793.1 - 754
AK170696.1 - 4 777
AK171042.1 - 4 786
AK171142.1 - 1541
AK172606.1 - 2 1436
AK186454.1 - 1 145
BC021547.1 - 4 780 4507218

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase II promoter
intracellular receptor signaling pathway
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
ligand-dependent nuclear receptor transcription coactivator activity
transcription regulatory region DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header