This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GGCX SIRNA Assays ›
Gene Symbol:
Ggcx
Gene Name:
gamma-glutamyl carboxylase
Gene Aliases:
-
Chromosome Location:
Chr.6: 72414296 - 72431106 on Build GRCm38
UniGene ID:
Mm.19937
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 185627

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ggcx
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019802.5 NP_062776.1 2 265
GenBank mRNA AF087938.1 - 2 240
AK033496.1 - 2 192
AK036577.1 - 2 239
AK135425.1 - 2 252
AK165982.1 - 2 240
BC138763.1 - 2 190 8861785
BC138764.1 - 2 190 8861786
BY310874.1 - 2 263

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

peptidyl-glutamic acid carboxylation
gamma-glutamyl carboxylase activity
lyase activity
vitamin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0