This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NDUFA1 SIRNA Assays ›
Gene Symbol:
Ndufa1
Gene Name:
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1
Gene Aliases:
1810049F12Rik, MWFE
Chromosome Location:
Chr.X: 37187588 - 37191238 on Build GRCm38
UniGene ID:
Mm.34869
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 185134

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ndufa1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019443.2 NP_062316.1 2 265
GenBank mRNA AK007840.1 - 2 172
AK131584.1 - 464
BC024364.1 - 2 174 5031206
CF383819.1 - 1 49
CT010398.1 - 2 117
Y07708.1 - 2 284

Back To Top

More Information


Gene Ontology Categories:

Function(s)

transport
oxidation-reduction process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0