This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD160 SIRNA Assays ›
Gene Symbol:
Cd160
Gene Name:
CD160 antigen
Gene Aliases:
AU045688, By55
Chromosome Location:
Chr.3: 96796883 - 96829351 on Build GRCm38
UniGene ID:
Mm.34693
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 184882

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cd160
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163496.1 NP_001156968.1 4 658
NM_018767.3 NP_061237.3 4 581
GenBank mRNA AF060982.1 - 4 552
AK042093.1 - 4 582
AK139789.1 - 4 659
AK145332.1 - 4 639
BC021596.1 - 4 553 5098004

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
defense response to Gram-negative bacterium
receptor binding
MHC class I receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header