This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other YAE1D1 SIRNA Assays ›
Gene Symbol:
Yae1d1
Gene Name:
Yae1 domain containing 1
Gene Aliases:
1600012F09Rik
Chromosome Location:
Chr.13: 17986640 - 17993351 on Build GRCm38
UniGene ID:
Mm.252918
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 183658

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Yae1d1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025904.3 NP_080180.1 1 120
GenBank mRNA AK005425.1 - 1 107
AK011774.1 - 1 119
AK011805.1 - 1 88
AK011852.1 - 1 88
AK019184.1 - 1 74
AK030455.1 - 1 100
AK076034.1 - 1 77
AK151981.1 - 1 98
BC020123.1 - 1 85
BC037189.1 - 1 107
BC048453.1 - 1 103 6595060
BC083111.1 - 1 79 5329860
CJ084672.1 - 1 120

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0