This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM170 SIRNA Assays ›
Gene Symbol:
Tmem170
Gene Name:
transmembrane protein 170
Gene Aliases:
9030409E16Rik, AA403674, D8Bwg1414e, Tmem170a
Chromosome Location:
Chr.8: 111863038 - 111876863 on Build GRCm38
UniGene ID:
Mm.345350
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 183425

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem170
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025781.2 NP_080057.1 2 273
GenBank mRNA AK007926.1 - 2 317
AK018496.1 - 2 274
BC139377.1 - 2 348 8862399
BC145964.1 - 2 348 40131267

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm