This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EMC2 SIRNA Assays ›
Gene Symbol:
Emc2
Gene Name:
ER membrane protein complex subunit 2
Gene Aliases:
4921531G14Rik, AV060620, AW209495, Ttc35
Chromosome Location:
Chr.15: 43477214 - 43527777 on Build GRCm38
UniGene ID:
Mm.244512
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 183339

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Emc2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025736.2 NP_080012.1 11 1167
GenBank mRNA AK014989.1 - 11 1168
AK015968.1 - 11 1133
AK172887.1 - 11 1112
BC004716.1 - 11 1125 3582196

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0