This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1700029F12RIK SIRNA Assays ›
Gene Symbol:
1700029F12Rik
Gene Name:
RIKEN cDNA 1700029F12 gene
Gene Aliases:
-
Chromosome Location:
Chr.13: 97021864 - 97035254 on Build GRCm38
UniGene ID:
Mm.390629
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 183079

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700029F12Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080777.2 NP_001074246.1 3 641
NM_001284205.1 NP_001271134.1 3 599
NM_025585.3 NP_079861.1 3 629
GenBank mRNA AK006036.1 - 3 617
AK006488.1 - 3 597
BC049576.1 - 3 615 6772947
BU962717.1 - 3 602
CF106098.1 - 2 451

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0