This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM218 SIRNA Assays ›
Gene Symbol:
Tmem218
Gene Name:
transmembrane protein 218
Gene Aliases:
1810021J13Rik, AV004931
Chromosome Location:
Chr.9: 37208223 - 37223872 on Build GRCm38
UniGene ID:
Mm.41451
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 182875

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem218
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025464.2 NP_079740.1 3 221
GenBank mRNA AK003010.1 - 3 218
AK007575.1 - 3 205
AK008902.1 - 3 192
AK161115.1 - 223
BC034160.1 - 3 221 5049687

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0