This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OTOR SIRNA Assays ›
Gene Symbol:
Otor
Gene Name:
otoraplin
Gene Aliases:
CDRAP, Fdp, MIA, MIAL
Chromosome Location:
Chr.2: 143078492 - 143081700 on Build GRCm38
UniGene ID:
Mm.157751
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 181684

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Otor
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020595.2 NP_065620.1 3 289
GenBank mRNA AF243504.1 - 3 289
AJ243939.1 - 3 304
AK047965.1 - 1 1478
BC139152.1 - 3 388 8862174
BC139153.1 - 3 388 8862175

Back To Top

More Information


Gene Ontology Categories:

Function(s)

cartilage condensation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0