This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RBM42 SIRNA Assays ›
Gene Symbol:
Rbm42
Gene Name:
RNA binding motif protein 42
Gene Aliases:
1700003D06Rik, 3100004P22Rik
Chromosome Location:
Chr.7: 30640995 - 30650228 on Build GRCm38
UniGene ID:
Mm.401747
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 181136

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rbm42
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133693.2 NP_598454.2 10 1523
GenBank mRNA AK013929.1 - 10 1464
AK151000.1 - 10 1435
BC009148.1 - 10 1460 3600406
BC011286.1 - 10 1442 3485410
BC027372.1 - 10 1447 4952304
BC039917.1 - 10 1458
BC057928.1 - 10 1449 5289543
BY551742.1 - 3 323

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001014159, XM_006228781, BC079321, FQ235070, FQ216075.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of mRNA splicing, via spliceosome
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0