This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM135A SIRNA Assays ›
Gene Symbol:
Fam135a
Gene Name:
family with sequence similarity 135, member A
Gene Aliases:
4921533L14Rik, AI256732, AI835491
Chromosome Location:
Chr.1: 24010758 - 24100460 on Build GRCm38
UniGene ID:
Mm.87130
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 180406

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam135a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026604.4 NP_080880.4 4 556
GenBank mRNA AK053786.1 - 4 475
AK081262.1 - 4 501
AK149303.1 - 4 548
AK165306.1 - 4 557
BC070446.1 - 3 238 30362784
BY732479.1 - 4 557

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001134597, XM_006244664, XM_006244665, XM_006244666, FQ222914.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0