This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD226 SIRNA Assays ›
Gene Symbol:
CD226
Gene Name:
CD226 molecule
Gene Aliases:
DNAM-1, DNAM1, PTA1, TLiSA1
Chromosome Location:
Chr.18: 69860228 - 69962086 on Build GRCh38
UniGene ID:
Hs.660130
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 18006

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CD226
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303618.1 NP_001290547.1 2 596
NM_006566.3 NP_006557.2 3 598
XM_005266642.3 XP_005266699.1 3 697
XM_017025524.1 XP_016881013.1 2 596
GenBank mRNA AJ537553.1 - 2 178
AK313199.1 - 2 597
BC074787.2 - 2 183 30915562
GU935335.1 - 3 345
U56102.1 - 3 357

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytokine production
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
positive regulation of immunoglobulin mediated immune response
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
signal transduction
cell recognition
positive regulation of mast cell activation
positive regulation of natural killer cell mediated cytotoxicity
regulation of immune response
positive regulation of Fc receptor mediated stimulatory signaling pathway
receptor activity
integrin binding
protein binding
protein kinase binding
protein homodimerization activity
cell adhesion molecule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0