This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF38 SIRNA Assays ›
Gene Symbol:
Rnf38
Gene Name:
ring finger protein 38
Gene Aliases:
1700065B19Rik, 2610202O07Rik, AA673263, Oip1
Chromosome Location:
Chr.4: 44126212 - 44233643 on Build GRCm38
UniGene ID:
Mm.262859
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 178911

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf38
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038993.3 NP_001034082.1 11 1459
NM_175201.5 NP_780410.2 10 1489
GenBank mRNA AK006888.1 - 9 1328
AK147608.1 - 11 1516
AK163346.1 - 6 620
BC060730.1 - 10 1466 6408623
BC062976.1 - 10 1403 6848803

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
ubiquitin-protein transferase activity
zinc ion binding
UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
ligase activity
ribosomal S6-glutamic acid ligase activity
coenzyme F420-0 gamma-glutamyl ligase activity
coenzyme F420-2 alpha-glutamyl ligase activity
metal ion binding
protein-glycine ligase activity
protein-glycine ligase activity, initiating
protein-glycine ligase activity, elongating
tubulin-glycine ligase activity
protein-glutamic acid ligase activity
tubulin-glutamic acid ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0