This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 4930506M07RIK SIRNA Assays ›
Gene Symbol:
4930506M07Rik
Gene Name:
RIKEN cDNA 4930506M07 gene
Gene Aliases:
Kiaa1598, Shootin1, mKIAA1598
Chromosome Location:
Chr.19: 58973358 - 59076122 on Build GRCm38
UniGene ID:
Mm.37338
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 178846

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4930506M07Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114312.1 NP_001107784.1 4, 5 818
NM_175172.4 NP_780381.1 4, 5 818
GenBank mRNA AK082304.1 - 4, 5 819
AK220405.1 - 4, 5 848
BC030338.1 - 4, 5 390 2599101
EF055486.1 - 4, 5 261

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
axonogenesis
neuron projection morphogenesis
membrane depolarization
regulation of establishment of cell polarity
regulation of neuron migration
protein binding
kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0