This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DPPA3 SIRNA Assays ›
Gene Symbol:
Dppa3
Gene Name:
developmental pluripotency-associated 3
Gene Aliases:
2410075G02Rik, PCG7, PGC7, Stella
Chromosome Location:
Chr.6: 122626424 - 122630271 on Build GRCm38
UniGene ID:
Mm.27982
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 178616

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dppa3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139218.1 NP_631964.1 2 207
GenBank mRNA AB072734.1 - 2 108
AF490347.1 - 2 212
AK010723.1 - 2 145
AK135769.1 - 2 212
AK136263.1 - 2 212
AY082485.1 - 2 207
AY134859.1 - 2 219
BC099433.1 - 2 185 30933569
BC100331.1 - 2 182 30934920
BC107340.1 - 2 139 40055218
BC107341.1 - 2 139 40055220

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001047864.

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
chromatin modification
embryonic cleavage
protection of DNA demethylation of female pronucleus
negative regulation of DNA demethylation
regulation of genetic imprinting
protein binding
methylated histone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0