This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PELP1 SIRNA Assays ›
Gene Symbol:
Pelp1
Gene Name:
proline, glutamic acid and leucine rich protein 1
Gene Aliases:
4930563C04Rik, MNAR
Chromosome Location:
Chr.11: 70392881 - 70410031 on Build GRCm38
UniGene ID:
Mm.340601
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 178257

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pelp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029231.4 NP_083507.3 1 146
GenBank mRNA AK005027.1 - 1 144
AK016197.1 - 1 147
AK128966.1 - 1 146
BC090620.1 - 1 140 30699968

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cellular response to estrogen stimulus
chromatin binding
transcription factor binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0