This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM180 SIRNA Assays ›
Gene Symbol:
Tmem180
Gene Name:
transmembrane protein 180
Gene Aliases:
4930449A08Rik, 4930538D17Rik
Chromosome Location:
Chr.19: 46356877 - 46375254 on Build GRCm38
UniGene ID:
Mm.159290
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 178250

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem180
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029186.2 NP_083462.2 4 817
GenBank mRNA AK015988.1 - 1 174
AK031554.1 - 4 763
AK151797.1 - 4 792
AK161305.1 - 4 773
AK170247.1 - 764
BC026385.1 - 2 945
BC031169.1 - 4 762
BC049142.1 - 1 103
BC058746.1 - 4 763 6399588

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0