This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DBNDD1 SIRNA Assays ›
Gene Symbol:
Dbndd1
Gene Name:
dysbindin (dystrobrevin binding protein 1) domain containing 1
Gene Aliases:
2810427I04Rik, D8Ertd590e
Chromosome Location:
Chr.8: 123504718 - 123518041 on Build GRCm38
UniGene ID:
Mm.27925
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 177473

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dbndd1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170975.2 NP_001164446.2 3 959
NM_028146.4 NP_082422.3 4 1100
GenBank mRNA AK013173.1 - 4 998
AK028187.1 - 4 1037
BC046622.1 - 3 894 6389902

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0