This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CNN3 SIRNA Assays ›
Gene Symbol:
Cnn3
Gene Name:
calponin 3, acidic
Gene Aliases:
1600014M03Rik, C85854, Calpo3
Chromosome Location:
Chr.3: 121426541 - 121458205 on Build GRCm38
UniGene ID:
Mm.275555
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 177353

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cnn3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028044.2 NP_082320.1 4 513
GenBank mRNA AK005460.1 - 4 515
AK051121.1 - 3 731
AK051628.1 - 4 531
AK081672.1 - 4 429
AK159555.1 - 4 558
AK159871.1 - 4 543
AK166578.1 - 4 521
AK167201.1 - 4 534
AK167613.1 - 4 531
AK169021.1 - 4 538
AK169098.1 - 4 558
AK169410.1 - 4 488
AK179383.1 - 4 244
AK180041.1 - 3 263
AK184231.1 - 4 249
AK190584.1 - 3 258
AK208457.1 - 3 213
AK212516.1 - 3 162
AK215736.1 - 3 223
AK216318.1 - 4 244
BC055711.1 - 4 514 6415164
BC085268.1 - 4 291 6487559

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

actomyosin structure organization
negative regulation of ATPase activity
actin binding
protein binding
calmodulin binding
microtubule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0