This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF215 SIRNA Assays ›
Gene Symbol:
Rnf215
Gene Name:
ring finger protein 215
Gene Aliases:
0610009J22Rik, AW557025, C77903
Chromosome Location:
Chr.11: 4135153 - 4141193 on Build GRCm38
UniGene ID:
Mm.240586
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 177157

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnf215
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027859.2 NP_082135.2 9 1305
GenBank mRNA AK002414.1 - 9 1263
AK047194.1 - 1254
AK154140.1 - 7 1207
BC103627.1 - 9 1229 6486672
BC115907.1 - 9 1252 40105152
BC115908.1 - 9 1252 40105148

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107234, XM_006251300.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
zinc ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0