siRNA Details
Gene Symbol: Tchhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027762.3 | NP_082038.2 | 3 | 788 | ||
GenBank mRNA | AA500450.1 | - | 1 | 18 | ||
AK017244.1 | - | 3 | 789 |

Gene Symbol: Tchhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027762.3 | NP_082038.2 | 3 | 788 | ||
GenBank mRNA | AA500450.1 | - | 1 | 18 | ||
AK017244.1 | - | 3 | 789 |