This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AOX3 SIRNA Assays ›
Gene Symbol:
Aox3
Gene Name:
aldehyde oxidase 3
Gene Aliases:
1200011D03Rik, AI326299, Aoh1, Aoh2
Chromosome Location:
Chr.1: 58113136 - 58200452 on Build GRCm38
UniGene ID:
Mm.20108
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 176712

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aox3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023617.2 NP_076106.2 35 4096
GenBank mRNA AF172276.2 - 35 4175
AK004700.1 - 24 2892
AK030109.1 - 31 3637
AK149434.1 - 35 4058
AY347569.1 - 35 3976
AY347570.1 - 35 3976
BC049853.1 - 35 4047
BC138876.1 - 35 4075 8861898

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001008527, XM_006245004, XM_008767121, XM_008767123, XM_008767122, and others.

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
catalytic activity
aldehyde oxidase activity
iron ion binding
UDP-N-acetylmuramate dehydrogenase activity
electron carrier activity
oxidoreductase activity
oxidoreductase activity, acting on CH-OH group of donors
molybdenum ion binding
metal ion binding
flavin adenine dinucleotide binding
NAD binding
iron-sulfur cluster binding
2 iron, 2 sulfur cluster binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0