This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1700028K03RIK SIRNA Assays ›
Gene Symbol:
1700028K03Rik
Gene Name:
RIKEN cDNA 1700028K03 gene
Gene Aliases:
-
Chromosome Location:
Chr.5: 107534711 - 107551542 on Build GRCm38
UniGene ID:
Mm.67037
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 176588

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700028K03Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175241.1 NP_780450.1 3 242
NM_182745.2 NP_877422.1 3 242
GenBank mRNA AK006463.1 - 3 243
AK029908.1 - 3 197
AK032886.1 - 4 442
CN835533.1 - 3 217

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0