This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM50B SIRNA Assays ›
Gene Symbol:
TMEM50B
Gene Name:
transmembrane protein 50B
Gene Aliases:
C21orf4, HCVP7TP3
Chromosome Location:
Chr.21: 33432486 - 33480009 on Build GRCh38
UniGene ID:
Hs.433668
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 17577

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM50B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006134.6 NP_006125.2 4 465
NR_040016.1 4 465
XM_011529746.1 XP_011528048.1 4 463
GenBank mRNA AF045606.2 - 4 410
AF086280.1 - 3 299
AK309977.1 - 4 392
AK310701.1 - 4 321
AK311870.1 - 3 285
AY358634.1 - 4 389
AY820138.1 - 3 252
BC000569.2 - 4 381 3342648
DA286739.1 - 4 466
DQ286229.1 - 3 252

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0