This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MRPL43 SIRNA Assays ›
Gene Symbol:
Mrpl43
Gene Name:
mitochondrial ribosomal protein L43
Gene Aliases:
4930442D21Rik, bMRP36a
Chromosome Location:
Chr.19: 45005014 - 45006442 on Build GRCm38
UniGene ID:
Mm.44174
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 175544

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mrpl43
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053164.3 NP_444394.1 3 692
GenBank mRNA AB049657.1 - 3 652
AK015357.1 - 3 673
AK076204.1 - 3 651
BC027676.1 - 3 632
BC048733.1 - 3 683 6512868
BC065812.1 - 3 639

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

translation
structural constituent of ribosome
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0