This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CWC22 SIRNA Assays ›
Gene Symbol:
Cwc22
Gene Name:
CWC22 spliceosome-associated protein homolog (S. cerevisiae)
Gene Aliases:
AA684037, AI173004, AL022752, B230213M24, mKIAA1604
Chromosome Location:
Chr.2: 77895653 - 77946356 on Build GRCm38
UniGene ID:
Mm.288151
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 174923

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cwc22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290740.1 NP_001277669.1 10 1381
NM_030560.5 NP_085037.2 11 1439
NM_172667.2 NP_766255.1 10 1378
GenBank mRNA AK045589.1 - 10 1210
AK077961.1 - 11 1267
AK089988.1 - 1217
AK129404.1 - 1195
AK132001.1 - 10 1322
AK149528.1 - 10 1205
AK166487.1 - 10 1217
BC003273.1 - 4 366
BC003993.1 - 10 1191 3492146

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0