This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNIP4 SIRNA Assays ›
Gene Symbol:
Kcnip4
Gene Name:
Kv channel interacting protein 4
Gene Aliases:
AV032399, Calp, Calp250, KChIP4, KChIP4a
Chromosome Location:
Chr.5: 48389503 - 49525122 on Build GRCm38
UniGene ID:
Mm.160172
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 174903

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnip4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199242.1 NP_001186171.1 2 306
GenBank mRNA BC051130.1 - 2 306 6532082
DQ148501.1 - 2 306

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_025221, DQ148487, BI596848, AK289922, AK314058, and others.
This siRNA also targets the following transcripts in Rat: NM_181365, AF345444, AF453245.

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
potassium ion transport
regulation of ion transmembrane transport
potassium ion transmembrane transport
protein localization to plasma membrane
regulation of potassium ion transmembrane transport
voltage-gated ion channel activity
potassium channel activity
calcium ion binding
potassium channel regulator activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0