This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PARP9 SIRNA Assays ›
Gene Symbol:
Parp9
Gene Name:
poly (ADP-ribose) polymerase family, member 9
Gene Aliases:
ARTD9, AW214463, BC003281, Bagl, Bal, PARP-9
Chromosome Location:
Chr.16: 35938470 - 35972621 on Build GRCm38
UniGene ID:
Mm.49074
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 174881

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Parp9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030253.3 NP_084529.1 4 975
GenBank mRNA AK037903.1 - 4 542
AK050032.1 - 4 419
BC003281.1 - 4 417 3581841
BC010312.1 - 3 400
BC070466.1 - 4 975 30615225

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA repair
double-strand break repair
cellular response to DNA damage stimulus
regulation of response to interferon-gamma
NAD+ ADP-ribosyltransferase activity
transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0