This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DOK5 SIRNA Assays ›
Gene Symbol:
Dok5
Gene Name:
docking protein 5
Gene Aliases:
2700055C10Rik
Chromosome Location:
Chr.2: 170731807 - 170879775 on Build GRCm38
UniGene ID:
Mm.41633
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 174572

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dok5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163686.1 NP_001157158.1 4 621
NM_029761.4 NP_084037.3 4 621
GenBank mRNA AF418208.1 - 4 483
AK012430.1 - 4 297
AK044148.1 - 4 489
BC066822.1 - 4 621 30021902

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
transmembrane receptor protein tyrosine kinase signaling pathway
nervous system development
intracellular signal transduction
regulation of neurotrophin TRK receptor signaling pathway
receptor signaling protein activity
insulin receptor binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0