This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRRT3 SIRNA Assays ›
Gene Symbol:
Prrt3
Gene Name:
proline-rich transmembrane protein 3
Gene Aliases:
6330505P20, B230206N24Rik
Chromosome Location:
Chr.6: 113493638 - 113502235 on Build GRCm38
UniGene ID:
Mm.133060
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 173617

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prrt3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289699.1 NP_001276628.1 2 754
NM_001289700.1 NP_001276629.1 2 675
NM_172487.4 NP_766075.2 2 728
GenBank mRNA AK031947.1 - 2 678
AK045491.1 - 2 632
AK045678.1 - 2 716
AK136854.1 - 2 708
BC058349.1 - 2 728 5695444

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header