This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HLCS SIRNA Assays ›
Gene Symbol:
Hlcs
Gene Name:
holocarboxylase synthetase (biotin- [propriony-Coenzyme A-carboxylase (ATP-hydrolysing)] ligase)
Gene Aliases:
410I21.SP6, D16Jhu34
Chromosome Location:
Chr.16: 94129195 - 94313905 on Build GRCm38
UniGene ID:
Mm.30921
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 173154

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hlcs
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139145.4 NP_631884.1 3 1041
GenBank mRNA AB066227.1 - 2 901
AK052424.1 - 4 1337
AK158212.1 - 4 1158
AK164351.1 - 4 1322
BC050090.1 - 3 1042 30012868

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
metabolic process
cell proliferation
protein biotinylation
histone modification
response to biotin
histone biotinylation
nucleotide binding
catalytic activity
biotin-[acetyl-CoA-carboxylase] ligase activity
biotin-[methylcrotonoyl-CoA-carboxylase] ligase activity
biotin-[methylmalonyl-CoA-carboxytransferase] ligase activity
biotin-[propionyl-CoA-carboxylase (ATP-hydrolyzing)] ligase activity
ATP binding
biotin binding
ligase activity
lipoate-protein ligase activity
biotin-protein ligase activity
enzyme binding
tRNA(Ile)-lysidine synthase activity
protein homodimerization activity
biotin-[pyruvate-carboxylase] ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0