This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other H1FOO SIRNA Assays ›
Gene Symbol:
H1foo
Gene Name:
H1 histone family, member O, oocyte-specific
Gene Aliases:
C86609, H1.8, H1fo, H1oo
Chromosome Location:
Chr.6: 115944294 - 115950242 on Build GRCm38
UniGene ID:
Mm.21899
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 173012

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: H1foo
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138311.2 NP_612184.1 2, 3 405
GenBank mRNA AK135944.1 - 2, 3 409
AK162137.1 - 2, 3 384
AK162154.1 - 2, 3 402
AK165127.1 - 3, 4 487
AY007195.1 - 2, 3 405
BC137916.1 - 2, 3 401 8860935
BC145046.1 - 2, 3 401

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
nucleosome positioning
regulation of DNA methylation
meiotic cell cycle
negative regulation of stem cell differentiation
DNA binding
nucleosomal DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0