This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SMIM3 SIRNA Assays ›
Gene Symbol:
Smim3
Gene Name:
small integral membrane protein 3
Gene Aliases:
2010002N04Rik, AI195350, AW561905, Nid67, cI-41
Chromosome Location:
Chr.18: 60474191 - 60518947 on Build GRCm38
UniGene ID:
Mm.273197
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 172894

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Smim3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134133.2 NP_598894.1 2 1653
GenBank mRNA AF313412.1 - 2 1195
AK143383.1 - 2 1657
AK155873.1 - 2 1299
AK168283.1 - 2 1476

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_173126, XM_006254765, XM_008772096, XM_006254769, XM_006254767, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0