This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDCP1 SIRNA Assays ›
Gene Symbol:
Cdcp1
Gene Name:
CUB domain containing protein 1
Gene Aliases:
9030022E12Rik, AA409659, E030027H19Rik
Chromosome Location:
Chr.9: 123170805 - 123216124 on Build GRCm38
UniGene ID:
Mm.27519
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 172775

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdcp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133974.3 NP_598735.2 2 326
GenBank mRNA AK035674.1 - 2 288
AK087108.1 - 2 224
BC018410.1 - 2 337
BC059937.1 - 2 233
BC085253.1 - 2 326 5347122

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0