This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C8B SIRNA Assays ›
Gene Symbol:
C8b
Gene Name:
complement component 8, beta polypeptide
Gene Aliases:
4930439B20Rik, AI595927
Chromosome Location:
Chr.4: 104764576 - 104804549 on Build GRCm38
UniGene ID:
Mm.87072
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 172769

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C8b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133882.2 NP_598643.1 2 270
GenBank mRNA AB077304.1 - 2 192
AB077305.1 - 2 192
AK050313.1 - 2 271
BC096382.1 - 2 244 5102825

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune system process
immune response
complement activation, alternative pathway
complement activation, classical pathway
cytolysis
innate immune response
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0