This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other USP33 SIRNA Assays ›
Gene Symbol:
Usp33
Gene Name:
ubiquitin specific peptidase 33
Gene Aliases:
9830169D19Rik, AA409780, Vdu1
Chromosome Location:
Chr.3: 152346478 - 152393617 on Build GRCm38
UniGene ID:
Mm.258320
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 172706

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Usp33
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001076676.2 NP_001070144.1 5 446
NM_001252486.1 NP_001239415.1 5 446
NM_133247.3 NP_573510.2 5 446
GenBank mRNA AF383174.1 - 5 435
AK047530.1 - 5 408
AK053227.1 - 5 397
AK082251.1 - 5 429
AK082943.1 - 5 429
AK087467.1 - 5 447
AK122442.1 - 4 277
AK143844.1 - 5 414
BC005506.1 - 5 399
BC067399.1 - 4 288
BC089315.1 - 5 392
BY731439.1 - 5 443

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
ubiquitin-dependent protein catabolic process
endocytosis
axon guidance
regulation of G-protein coupled receptor protein signaling pathway
cellular response to starvation
regulation of autophagy
cell migration
protein deubiquitination
negative regulation of protein binding
positive regulation of protein binding
proteasome-mediated ubiquitin-dependent protein catabolic process
protein stabilization
centrosome duplication
regulation of proteasomal protein catabolic process
protein K63-linked deubiquitination
protein K48-linked deubiquitination
G-protein coupled receptor binding
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
peptidase activity
cysteine-type peptidase activity
zinc ion binding
hydrolase activity
Ral GTPase binding
thiol-dependent ubiquitinyl hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0