This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SCML2 SIRNA Assays ›
Gene Symbol:
Scml2
Gene Name:
sex comb on midleg-like 2 (Drosophila)
Gene Aliases:
4932420G07Rik
Chromosome Location:
Chr.X: 161082629 - 161258218 on Build GRCm38
UniGene ID:
Mm.159173
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 172615

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Scml2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290651.1 NP_001277580.1 6 1000
NM_001290652.1 NP_001277581.1 6 1000
GenBank mRNA AF285577.1 - 8 826
AK016533.1 - 9 872
AK031490.1 - 6 979
AK040521.1 - 6 1002
BC056382.1 - 6 690 6853936

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0