This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LIMS1 SIRNA Assays ›
Gene Symbol:
Lims1
Gene Name:
LIM and senescent cell antigen-like domains 1
Gene Aliases:
2310016J22Rik, 4921524A02Rik, AI507642, AU021743, AW551584, C430041B13Rik, Lims1l, PINCH, PINCH1
Chromosome Location:
Chr.10: 58323435 - 58424691 on Build GRCm38
UniGene ID:
Mm.402697
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 172029

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lims1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193303.1 NP_001180232.1 2 336
NM_026148.3 NP_080424.2 2 330
NM_201242.1 NP_957694.1 2 354
GenBank mRNA AK009390.1 - 2 337
AK014954.1 - 2 331
AK049564.1 - 1 670
AK075667.1 - 1 103
AK082932.1 - 2 199
AK088939.1 - 2 205
AK151235.1 - 2 205
AK151938.1 - 2 197
AK152679.1 - 2 197
AK153346.1 - 2 205
BC005621.1 - 2 185 3711611
BC067393.1 - 2 128
DQ003608.1 - 2 321
EU122953.1 - 2 320
EU122954.1 - 2 188

Back To Top

More Information


Panther Classification:

Molecular Function -

cell junction protein

Gene Ontology Categories:

Function(s) Process(es)

cell-matrix adhesion
establishment or maintenance of cell polarity
positive regulation of gene expression
positive regulation of cell-substrate adhesion
single organismal cell-cell adhesion
chordate embryonic development
negative regulation of apoptotic process
positive regulation of GTPase activity
establishment of protein localization
cell-cell junction organization
negative regulation of transcription, DNA-templated
negative regulation of epithelial cell proliferation
protein heterooligomerization
positive regulation of focal adhesion assembly
positive regulation of substrate adhesion-dependent cell spreading
negative regulation of neural precursor cell proliferation
negative regulation of hepatocyte proliferation
positive regulation of integrin-mediated signaling pathway
protein binding
zinc ion binding
protein kinase binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0