This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC28A3 SIRNA Assays ›
Gene Symbol:
Slc28a3
Gene Name:
solute carrier family 28 (sodium-coupled nucleoside transporter), member 3
Gene Aliases:
Cnt3
Chromosome Location:
Chr.13: 58553307 - 58610877 on Build GRCm38
UniGene ID:
Mm.18188
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 171928

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc28a3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022317.3 NP_071712.3 8 966
GenBank mRNA AF305211.1 - 8 899
AK052784.1 - 8 967
AK145077.1 - 8 967
BC010472.1 - 8 954 3586005
BC013783.1 - 8 957 3600786

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_022127, NM_001199633, AK304406, AK310861, AF305210, and others.
This siRNA also targets the following transcripts in Rat: NM_080908, XM_008771422, AY059414.

Gene Ontology Categories:

Function(s) Process(es)

retina homeostasis
transport
purine nucleobase transport
pyrimidine nucleobase transport
purine nucleoside transmembrane transport
pyrimidine nucleoside transport
sodium ion transmembrane transport
pyrimidine-containing compound transmembrane transport
nucleoside transmembrane transport
nucleoside binding
nucleoside transmembrane transporter activity
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
purine-specific nucleoside:sodium symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0