This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MED16 SIRNA Assays ›
Gene Symbol:
Med16
Gene Name:
mediator complex subunit 16
Gene Aliases:
95kDa, A630083L04, Thrap5, Trap95
Chromosome Location:
Chr.10: 79894707 - 79908941 on Build GRCm38
UniGene ID:
Mm.260089
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 171799

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Med16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163276.1 NP_001156748.1 5 645
NM_198107.2 NP_932775.2 5 648
GenBank mRNA AK042340.1 - 5 610
AK149345.1 - 5 629
BC057056.1 - 5 602 6813010

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106768, XM_006240864, XM_006240862, XM_006240861, XM_006240865, and others.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
intracellular steroid hormone receptor signaling pathway
androgen receptor signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of receptor activity
transcription cofactor activity
catalytic activity
receptor activity
thyroid hormone receptor coactivator activity
thyroid hormone receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0