This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CEP120 SIRNA Assays ›
Gene Symbol:
Cep120
Gene Name:
centrosomal protein 120
Gene Aliases:
A230075C01, AU016693, Ccdc100
Chromosome Location:
Chr.18: 53681723 - 53745603 on Build GRCm38
UniGene ID:
Mm.193678
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 171625

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cep120
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178686.3 NP_848801.2 5 502
GenBank mRNA AK038916.1 - 5 564
AK044808.1 - 5 519
AK044952.1 - 5 496
AK047594.1 - 5 547
AK079319.1 - 5 519
AK089236.1 - 5 509
AK089242.1 - 5 509
BC053439.1 - 5 502 6328829

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
cell proliferation
cerebral cortex development
neurogenesis
interkinetic nuclear migration
astral microtubule organization
regulation of protein localization
regulation of microtubule-based process
protein binding
protein C-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0