This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBXO2 SIRNA Assays ›
Gene Symbol:
Fbxo2
Gene Name:
F-box protein 2
Gene Aliases:
FBG1, FBX2, Fbs1, Fbs2, NFB42, Prpl4
Chromosome Location:
Chr.4: 148160226 - 148166417 on Build GRCm38
UniGene ID:
Mm.262287
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 171420

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbxo2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176848.1 NP_789818.1 6 980
GenBank mRNA AK140175.1 - 6 1024
BC027053.1 - 5 933
BC046586.1 - 6 980 6487759

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
glycoprotein catabolic process
negative regulation of cell proliferation
protein ubiquitination
ER-associated ubiquitin-dependent protein catabolic process
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
regulation of protein ubiquitination
beta-amyloid binding
glycoprotein binding
protein binding
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0