This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCTD3 SIRNA Assays ›
Gene Symbol:
Kctd3
Gene Name:
potassium channel tetramerisation domain containing 3
Gene Aliases:
4930438A20Rik, 9330185B06, E330032J19Rik, NY-REN-45
Chromosome Location:
Chr.1: 188971095 - 189007868 on Build GRCm38
UniGene ID:
Mm.209880
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 170993

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kctd3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172650.2 NP_766238.1 18 3567
GenBank mRNA AK034380.1 - 18 3568
BC045196.1 - 5 1646
BC061458.1 - 18 3806
BC079581.1 - 18 3554 5702630

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein homooligomerization
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0