This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 1700021K19RIK SIRNA Assays ›
Gene Symbol:
1700021K19Rik
Gene Name:
RIKEN cDNA 1700021K19 gene
Gene Aliases:
5330403K09, Kiaa0226, Rubicon, mKIAA0226
Chromosome Location:
Chr.16: 32821702 - 32877348 on Build GRCm38
UniGene ID:
Mm.327319
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 170932

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700021K19Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001200038.1 NP_001186967.1 6 912
NM_172615.4 NP_766203.1 6 912
GenBank mRNA AK030368.1 - 6 886
AK042428.1 - 6 850
AK122221.1 - 6 876
AK170002.1 - 6 892
AK170223.1 - 6 850
AK170312.1 - 6 1143
AK171583.1 - 6 896
BC057307.1 - 6 816
BC060601.1 - 6 885 5706123
BC067390.1 - 6 897 30432960

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

endocytosis
autophagy
negative regulation of autophagy
negative regulation of phosphatidylinositol 3-kinase activity
negative regulation of endocytosis
negative regulation of autophagosome maturation
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0